Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2b Mouse anti-Rat, Biotin, Clone: G15-337, BD

Catalog No. BDB550327 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550327 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550327 Supplier BD Biosciences Supplier No. 550327
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G15-337 antibody reacts specifically with rat IgG2b. It does not react with other Ig isotypes. A suspension of pooled rat Ig was used as the source of immunogen.

Host Species: Mouse
Clone: G15-337
Isotype: IgG2b κ
Species Reactivity: Rat
Immunogen: Pooled rat Ig

Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2b
Applications ELISA
Classification Monoclonal
Clone G15-337
Concentration 250μg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing BSA, protein stabilizer, goat serum, and ≤ 0.09% sodium azide.
Host Species Mouse
Immunogen Pooled rat Ig
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.