Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

immunoglobulin kappa constant, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-615 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-615 Supplier Abnova Corporation Supplier No. H00003514B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human IGKC protein.

Sequence: MEAPAQLLFLLLLWLPDTTGEIVLTQSPVFLSLSPGEGATLSCRASQSVSRNLAWYQQKSGQAPRLLIYDASNRATGIPARFSGSGSGTDFTLTISSLEPEDSAVYYCQQRSNCLITFGQGTRLEIRRTVAAPSVFIFPPSDEQLKSGTASVVCLLNNFYPREAKVQWKVDNALQSGNSQESVTEQDSKDSTYSLSSTLTLSKADYEKHKVYACEVTHQGLSSPVTKSFNRGEC

Specifications

Antigen immunoglobulin kappa constant
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human IGKC protein.
Formulation No additive
Gene IGKC
Gene Accession No. BC066343.1
Gene Alias HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809
Gene Symbols IGKC
Host Species Mouse
Immunogen IGKC (AAH66343.1, 1 a.a. ∼ 234 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3514
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.