Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Mouse anti-Human, BB515, Clone: G20-127, BD

Catalog No. BDB564622 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564622 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564622 Supplier BD Biosciences Supplier No. 564622
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G20-127 monoclonal antibody specifically binds to the heavy chain of human IgM. The majority of mature B lymphocytes express IgM. The G20-127 antibody does not react with other immunoglobulin heavy chain isotypes.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM
Applications Flow Cytometry
Classification Monoclonal
Clone G20-127
Conjugate Brilliant Blue 515 (BB515)
Description IGHM; MU; Ig mu chain C region; AGM1; VH
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.