Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Mouse anti-Human, Brilliant Violet 605, Clone: G20-127, BD

Catalog No. BDB562977 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562977 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562977 Supplier BD Biosciences Supplier No. 562977
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G20-127 monoclonal antibody specifically binds to the heavy chain of human IgM. The majority of mature B lymphocytes express IgM. The G20-127 antibody does not react with other immunoglobulin heavy chain isotypes.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon™ BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon™ BV605 is a tandem fluorochrome of BD Horizon™ BV421 and Cy™3.5. Due to the excitation of Cy3.5 by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon™ PE-CF594 detectors off the green or yellow-green lasers. BD Horizon™ BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM
Applications Flow Cytometry
Classification Monoclonal
Clone G20-127
Conjugate Brilliant Violet 605
Description IGHM; MU; Ig mu chain C region; VH
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.