Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Rat anti-Mouse, HRP, Clone: II/41, BD

Catalog No. BDB550588 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550588 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550588 Supplier BD Biosciences Supplier No. 550588
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The II/41 clone has been reported to react specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. In addition, the II/41 clone has been reported not to stimulate B-cell proliferation.

Host Species: Rat
Clone: II/41
Isotype: IgG2a κ
Species Reactivity: Mouse

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM
Applications ELISA
Classification Monoclonal
Clone II/41
Conjugate HRP
Formulation Aqueous buffered solution containing BSA and ProClin™150 as preservative.
Host Species Rat
Immunogen Not reported
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.