Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Mouse anti-Human, PE-CF594, Clone: G20-127, BD

Catalog No. BDB562539 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562539 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562539 Supplier BD Biosciences Supplier No. 562539
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Antibody was conjugated with orizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. Conjugated to Horizon PE-CF594, developed as a better alternative to PE-Texas Red which excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics.

Host Species: Mouse
Clone: G20-127
Isotype: IgG1κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM
Applications Flow Cytometry
Classification Monoclonal
Clone G20-127
Concentration 5μL
Conjugate PE-CF594
Description IGHM; MU; Ig mu chain C region; VH
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.