Learn More
Description
The G20-127 monoclonal antibody specifically binds to the heavy chain of human IgM. The majority of mature B lymphocytes express IgM. The G20-127 antibody does not react with other immunoglobulin heavy chain isotypes.
Specifications
Specifications
| Antigen | IgM |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | G20-127 |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.