Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indian hedgehog homolog (Drosophila), Mouse, Clone: 2G9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant IHH.
Supplier: Abnova Corporation H00003549M01
Description
Sequence: ESGARVALSAVRPGDRVLAMGEDGSPTFSDVLIFLDREPHRLRAFQVIETQDPPRRLALTPAHLLFTADNHTEPAARFRATFASHVQPGQYVLVAGVPGSpecifications
Indian hedgehog homolog (Drosophila) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC034757 | |
IHH | |
IHH (AAH34757, 119 a.a. ∼ 217 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
3549 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Western Blot | |
2G9 | |
Mouse monoclonal antibody raised against a full length recombinant IHH. | |
IHH | |
BDA1/HHG2 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction