Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkBa Mouse anti-Human, Alexa Fluor 647, Clone: 25/IkBa/MAD-3, BD

Catalog No. BDB560817 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560817 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560817 Supplier BD Biosciences Supplier No. 560817
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NF-κB is a transcription factor that is a member of the mammalian NF-κB/Rel family of proteins. Members of this family are involved in the regulation of cell proliferation, immune function, as well as development.In resting cells, IκBα binds to and maintains NF-κB in the cytoplasm by blocking the nuclear localization sequences of NF-κB. In the cellular response to an extracellular signal, IκBα is phosphorylated and subsequently degraded via the ubiquination-proteasome pathway, allowing NF-κB to translocate to the nucleus. Once in the nucleus, NF-κB can induce the transcription of IκBα thereby renewing the cycle so that IκBα can form a complex with NF-κB and maintain it in its cytoplasmic location. IκBα -/- mice show an increased level of NF-κB activity and have been shown to die soon after birth.

The 25/IkBa/MAD-3 monoclonal antibody recognizes human IκBα regardless of phosphorylation status and does not cross-react with mouse IκBα .

TRUSTED_SUSTAINABILITY

Specifications

Antigen IkBa
Applications Flow Cytometry
Classification Monoclonal
Clone 25/IkBa/MAD-3
Conjugate Alexa Fluor 647
Description MAD-3; I-kappa-B-alpha; IkB-alpha; NFKBIA; NF-kappa-B inhibitor alpha
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IκBα aa. 145-302 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.