Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-IKK beta, Clone: Y466, Rabbit Monoclonal,

Catalog No. 04366MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-366-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-366-MI Supplier MilliporeSigma™ Supplier No. 04366

Specifications

Antigen IKK beta
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Y466
Conjugate Unconjugated
Formulation Rabbit Monoclonal in buffer containing glycerol, BSA, and sodium azide.
Gene Accession No. O14920
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human IKK beta.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NFKBIKB, IKK-beta, IKK2, IKKB, IKK-B, EC 2.7.11.10, FLJ33771, FLJ36218, FLJ38368, FLJ40509, MGC131801, OTTHUMP00000225854, OTTHUMP00000225858, OTTHUMP00000225860, OTTHUMP00000225865, OTTHUMP00000225866, IkBKB, EC 2.7.11
Target Species Human, Mouse
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.