Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKKΒ Mouse anti-Human, Rat, Mouse, Clone: 10AG2, MilliporeSigma™

Catalog No. 05535MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-535-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-535-MI Supplier MilliporeSigma Supplier No. 05535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IKKβ Antibody, clone 10AG2 detects level of IKKβ and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKKβ
Applications Western Blot
Classification Monoclonal
Clone 10AG2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing PBS containing 0.05% BSA, with 0.05% sodium azide. Frozen solution.
Gene Accession No. NM_001556
Gene Symbols IKBKB, IKK-beta, NFKBIKB, IKK2, FLJ40509, IKKB, MGC131801, IKK-B, IkBKB
Host Species Mouse
Immunogen His-tagged, full length human IKKβ/IKK2. Clone: 10AG2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.