Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKKγ Mouse, Unlabeled, Clone: C73-764, BD

Catalog No. BDB559675 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559675 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559675 Supplier BD Biosciences Supplier No. 559675
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The transcription factor NF-κB, (nuclear factor kappa-B) is controlled by interaction with an inhibitory subunit, IκB, which restricts NF-κB to the cytoplasm. Following stimulation by various cytokines or other stimuli, IκB becomes degraded and NF-κB is released to the nucleus. The release of IκB from NF-κB is a critical step in the activation of NF-κB signal pathways. A group of proteins form an NF-κB regulatory complex, or signalsome. Two members of this complex are a pair of closely related serine/threonine kinases, IKKα and IKKβ (aka IKK-1 and IKK-2), which phosphorylate critical residues of IκB, thus targeting it for subsequent degradation. The IKK complex contains similar amounts of IKKα, IKKβ, as well as two other polypeptides, which are differentially processed forms of a third subunit, IKKγ. IKKα and IKKβ become activated following phosphorylation by upstream kinases, including NF-κB-inducing kinase (NIK) and MEKK1. IKKγ interacts preferentially with IKKβ and is required for the activation of the IKK complex. Thus the IKK kinases play an important role in the activation of NF-κB. IKKγ migrates as a doublet between 50-55kDa in SDS-PAGE.

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IKKγ
Applications Western Blot
Classification Monoclonal
Clone C73-764
Concentration 0.5mg/mL
Conjugate Unconjugated
Description NEMO
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human IKKγ
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.