Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKKγ Mouse anti-Human, Mouse, MilliporeSigma™

Catalog No. 05631MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-631-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-631-MI Supplier MilliporeSigma Supplier No. 05631
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IKKγ Antibody is an antibody against IKKγ for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKKγ
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation PBS, pH 7.4, containing 0.02% sodium azide
Gene Accession No. NM_003639
Gene Symbols IKBKG, FIP3, IPD2, NEMO, IP2, Fip3p, IP1, IKKG, IKK-gamma, AMCBX1, FIP-3, IKKAP1, IP
Host Species Mouse
Immunogen His-tagged, full-length human IKKgamma/IKK3
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.