Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKKΕ Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07580MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-580-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-580-MI Supplier MilliporeSigma Supplier No. 07580
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-IKKΕ Antibody is validated for use in western blotting for the detection of IKKΕ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKKε
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation of 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols IKBKE; IKKI; MGC125294; IKK-i; IKKE; IKK-epsilon; IkBKE; MGC125295; MGC125297; KIAA0151
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 582-596 (SHLAKRLLQVFQEEC) of human IKKε
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_014002
Target Species Human, Mouse
Content And Storage −20°C for up to 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.