Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKKγ (pS376) Mouse anti-Human, PE, Clone: N19-39, BD

Catalog No. BDB562590 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562590 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562590 Supplier BD Biosciences Supplier No. 562590
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The N19-39 monoclonal antibody specifically binds to the IKK-gamma protein phosphorylated at the Ser376 site, ie, IKKγ (pS376). IKKγ is also known an NEMO (NF-κ-B essential modifier) and is encoded by the IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma) gene. The nuclear transcription factor κ-B, NF-κB, is controlled by interaction with an inhibitory subunit, IκB, which restricts NF-κB to the cytoplasm. Following stimulation by various cytokines or other stimuli, IκB becomes degraded and NF-κB is released to the nucleus. The release of IκB from NF-κB is thought to be a critical point in the activation of NF-κB signal pathways. Activated NF-κB regulates genes involved in various pathways including inflammation, immunity, and cell survival signaling pathways. A group of proteins form an NF-κB regulatory complex, or signalsome. Two members of this complex are a pair of closely related serine/threonine kinases, IKKα and IKKβ (also called IKK-1 and IKK-2), which phosphorylate critical residues of IκB, thus targeting it for subsequent degradation. The IKK complex contains similar amounts of IKKα, IKKβ as well as two other polypeptides, which are differentially processed forms of a third subunit, IKKγ. IKKα and IKKβ become activated following phosphorylation by upstream kinases, including NF-κB-inducing kinase (NIK)4 and MEKK1. IKKβ has been shown to phosphorylate IKKγ at serine 376 in response to signaling through the TNF receptor or the Tax oncoprotein of human T-cell leukemia virus type 1. IKKγ has an essential regulatory role in the IKK complex and is required for the activation of the NF-κB pathway in response to multiple cellular stimuli. IKKγ isoforms migrate as a doublet of ∽50-55kDa upon Western blot analysis.

Host Species: Mouse
Clone: N19-39
Isotype: IgG1, κ
Species Reactivity: Human
Immunogen: Phosphorylated Human IKKγ Peptide

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IKKγ (pS376)
Applications Flow Cytometry
Classification Monoclonal
Clone N19-39
Conjugate PE
Description IKBKG; IKKγ; IKKG; IKK-gamma; AMCBX1; FIP3; IKKAP1; IP1; IP2; IPD2; NEMO
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human IKKγ Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.