Learn More
IKKβ w/control Mouse anti-Human, Unlabeled, Clone: F18-1875, BD
Mouse Monoclonal Antibody with Control
Supplier: BD Biosciences 551920
Description
HeLa Control Lysate. Nuclear factor kappa B (NF-κB) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. The nuclear transcription factor kappa-B, NF-κB, is controlled by interaction with an inhibitory subunit, I-κB, which restricts NF-κB to the cytoplasm. Following stimulation by various cytokines or other stimuli, I-κB becomes degraded and NF-κB is released to the nucleus. The release of I-κB from NF-κB is thought to be a critical point in the activation of NF-κB signal pathways. A group of proteins form an NF-κB regulatory complex, or signalsome. Two members of this complex are a pair of closely related serine/threonine kinases, IKKα and IKKβ (also called IKK-1 and IKK-2), which phosphorylate critical residues of I-κB, thus targeting it for subsequent degradation. The IKK complex contains similar amounts of IKKα and IKKβ, as well as two other polypeptides, which are differentially processed forms of a third subunit, IKKγ. IKKα and IKKβ become activated following phosphorylation by upstream kinases, including NF-κB-inducing kinase (NIK) and MEKK1. IKKγ interacts preferentially with IKKβ and is required for the activation of the IKK complex. Experiments done on mice lacking IKKβ demonstrate that these mice died at mid-gestation from uncontrolled liver apoptosis. Furthermore, when cells from IKKβ-deficient m ice were treated with tumor necrosis factor α or interleukin 1, neither IKK nor NF-κB were activated. Thus IKKβ, but not IKKα, plays a pivotal role in the induction of NF-κB and activation of IKK. IKKβ migrates at ∽87kDa in SDS/PAGE. The antibody
recognizes human IKKβ. A recombinant protein corresponding to amino acids 1-666 of human IKKβ was used as the immunogen.
Host Species: Mouse
Clone: F18-1875
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IKKβ recombinant protein aa. 1-666
Western Blotting

Specifications
IKKβ w/control | |
Monoclonal | |
0.25mg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human IKKβ recombinant protein aa. 1-666 | |
50 μg | |
Cell Biology | |
Human | |
IgG1 |
Western Blot | |
F18-1875 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.