Learn More
IL-1β Hamster anti-Mouse, Unlabeled, Clone: B122, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 550605
Description
The B122 antibody recognizes the precursor and mature secreted forms of the mouse IL-1β protein. B122 neutralizes the biological activity of mouse IL-1β, but not the biological activities of mouse IL-1α nor human IL-1β. B122 does not recogn
Host Species: Hamster
Clone: B122
Isotype: IgG1 λ1
Species Reactivity: Mouse
Immunogen: Mouse IL-1β
ELISA, Immunoprecipitation, Western Blotting

Specifications
IL-1β | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse IL-1β | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 |
ELISA | |
B122 | |
Unconjugated | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.