Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 Β Rabbit anti-Pig, Polyclonal, MilliporeSigma™

Catalog No. AB10626MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10626MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10626MI Supplier MilliporeSigma Supplier No. AB10626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-IL-1 β Antibody is an antibody against IL-1 β for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 β
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antibodies in phosphate buffered saline, pH 7.2.
Gene Symbols IL1B; IL1F2; pro-interleukin-1-beta; IL1-BETA; Catabolin; IL-1; IL-1B; catabolin
Host Species Rabbit
Immunogen Recombinant swine IL-1β.
Purification Method Protein G purfied
Quantity 500 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000576.2
Target Species Pig
Content And Storage −20°C in undiluted aliquotes for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.