Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1α Hamster anti-Mouse, PE, Clone: ALF-161, BD

Catalog No. BDB559810 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559810 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559810 Supplier BD Biosciences Supplier No. 559810
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

This antibody recognizes the precursor, secreted and membrane-associated forms of mouse interleukin-1α (IL-1α) protein. No cross-reactivity was detected with mouse IL-1β. This antibody does not recognize human IL-1α or IL-1β. The cross-reactivity of this antibody with IL-1α from other species has not been tested. The immunogen used to generate this ALF-161 hybridoma was purified, recombinant mouse IL-1α protein. This is a neutralizing antibody.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1α
Applications Flow Cytometry
Classification Monoclonal
Clone ALF-161
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse IL-1α recombinant protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.