Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1 RI Fluorescein-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$197.90 - $397.50
Specifications
Antigen | IL-1 RI |
---|---|
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | Fluorescein |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB269F025
![]() |
R&D Systems
FAB269F025 |
25 Tests |
Each for $197.90
|
|
|||||
FAB269F100
![]() |
R&D Systems
FAB269F100 |
100 Tests |
Each for $397.50
|
|
|||||
Description
IL-1 RI Polyclonal specifically detects IL-1 RI in Human samples. It is validated for Flow Cytometry.Specifications
IL-1 RI | |
Flow Cytometry | |
Fluorescein | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
3554 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RI | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry 10 uL/10^6 cells | |
Polyclonal | |
Goat | |
Human | |
CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
IL1R1 | |
IgG | |
Affinity Purified | |
Detects human IL-1 RI in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title