Learn More
IL-10 Rat anti-Mouse, Biotin, Clone: SXC-1, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 554423
Description
The SXC-1 antibody reacts with mouse interleukin-10 (IL-10). The immunogen used to generate the SXC-1 hybridoma was mouse IL-10. This is a neutralizing antibody.
Host Species: Rat
Clone: SXC-1
Isotype: IgM
Species Reactivity: Mouse
Immunogen: Mouse IL-10 Protein
ELISA

Specifications
IL-10 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse IL-10 Protein | |
0.5 mg | |
Immunology | |
Mouse | |
IgM |
ELISA | |
SXC-1 | |
Biotin | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.