Learn More
IL-10 Rat anti-Human, Unlabeled, Clone: JES3-19F1, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 554705
Description
The JES3-19F1 antibody reacts with human (IL-10). The immunogen used to generate the JES3-19F1 hybridoma was recombinant human IL-10 expressed in COS cells. This is a neutralizing antibody. This antibody also reacts with viral IL-10.
Host Species: Rat
Clone: JES3-19F1
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Recombinant Human IL-10
Functional Assay, Neutralization
Specifications
| IL-10 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IL-10 | |
| 0.5 mg | |
| Immunology | |
| Human, Virus | |
| IgG2a |
| ELISA | |
| JES3-19F1 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.