Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Rat anti-Human, NA/LE, Unlabeled, Clone: JES3-19F1, BD

Catalog No. BDB554703 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554703 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554703 Supplier BD Biosciences Supplier No. 554703
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications ELISA
Classification Monoclonal
Clone JES3-19F1
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Interleukin-10; CSIF; Cytokine synthesis inhibitory factor; TGIF
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Immunogen Recombinant Human IL-10
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.