Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Rat anti-Mouse, Unlabeled, Clone: JES5-16E3, BD

Catalog No. BDB559063 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559063 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559063 Supplier BD Biosciences Supplier No. 559063
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). The immunogen used to generate the JES5-16E3 hybridoma was recombinant mouse IL-10. This is a neutralizing antibody. This antibody is routinely tested by immunocytochemistry. Other applications were tested at BD Biosciences Pharmingen during antibody development or reported in the literature.

Immunocytochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications Immunocytochemistry
Classification Monoclonal
Clone JES5-16E3
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant mouse IL-10
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.