Learn More
Description
Host Species: Rat
Clone JES5-16E3
Isotype IgG2b
Species Reactivity Ms
Immunogen Recombinant mouse IL-10
Flow Cytometry, Intracellular Staining
Specifications
Specifications
| Antigen | IL-10 |
| Classification | Monoclonal |
| Clone | JES5-16E3 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 510 |
| Host Species | Rat |
| Immunogen | Recombinant mouse IL-10 |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.