Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 (p40) Mouse anti-Human, Unlabeled, Clone: C8.3, BD

Catalog No. BDB551227 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551227 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551227 Supplier BD Biosciences Supplier No. 551227
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C8.3 antibody reacts with human IL-12 p40 subunit but not IL-12 p70 heterodimer or IL-12 p35 monomer. The immunogen used to generate the C8.3 hybridoma was the purified p40 subunit of human IL-12. The C8.3 antibody has been reported not to bind p40 chain when it is complexed to p35 (i.e., the p70 heterodimer). The combination of anti-p40 antibodies, C8.3 and C8.6 , has been reported to yield an assay for detecting only free p40 chain, and specifically not the p70 heterodimer. It has been reported that activated PBMC produce an excess of the
free p40 protein over the p70 heterodimer.

Host Species: Mouse
Clone: C8.3
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified human IL-12 p40 subunit

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 (p40)
Applications ELISA
Classification Monoclonal
Clone C8.3
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified human IL-12 p40 subunit
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.