Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 (p40/p70) Mouse anti-Human, Biotin, Clone: C8.6, BD

Catalog No. BDB554660 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554660 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554660 Supplier BD Biosciences Supplier No. 554660
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C8.6 antibody reacts with human IL-12 p40 monomers, homodimers, and p70 heterodimers, but not p35 subunits. The immunogen used to generate the C8.6 hybridoma was the purified p40 subunit of human IL-12. The C8.6 antibody is a neutralizing antibody and has been reported to strongly inhibit three different biological activities of IL-12 (i.e., IFN-γ induction), mitogenic effects on PHA blasts, and enhancement of NK cell-mediated cytotoxicity. C8.6 has been shown to immunoprecipitate IL-23. This antibody is routinely tested by ELISA as a detection antibody. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: C8.6
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified p40 Subunit of Human IL-12

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 (p40/p70)
Applications ELISA
Classification Monoclonal
Clone C8.6
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified p40 Subunit of Human IL-12
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.