Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 (p40/p70) Mouse anti-Human, Brilliant Violet 421, Clone: C8.6, BD

Catalog No. BDB565023 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565023 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565023 Supplier BD Biosciences Supplier No. 565023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C8.6 antibody reacts with human IL-12 p40 monomers, homodimers, and p70 heterodimers, but not p35 subunits. The immunogen used to generate the C8.6 hybridoma was the purified p40 subunit of human IL-12. The C8.6 antibody is a neutralizing antibody and has been reported to strongly inhibit three different biological activities of IL-12 (i.e., IFN- γinduction), mitogenic effects on PHA blasts, and enhancement of NK cell-mediated cytotoxicity. C8.6 has been shown to immunoprecipitate IL-23.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 (p40/p70)
Applications Flow Cytometry
Classification Monoclonal
Clone C8.6
Conjugate Brilliant Violet 421
Description IL12B; IL-12B; IL12p40; IL-12-40; CLMF; CLMF2; NKSF; NKSF2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Mouse
Immunogen Purified p40 Subunit of Human IL-12
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.