Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 (p40/p70) Mouse anti-Human, Unlabeled, Clone: C11.5, BD

Catalog No. BDB554573 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554573 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554573 Supplier BD Biosciences Supplier No. 554573
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C11.5 antibody reacts with human IL-12 p40 monomer and p70 heterodimer, but not p35 monomer. The immunogen used to generate the C11.5 hybridoma was the CHO-expressed recombinant human IL-12 p70 heterodimer. p40 has also been described as a subunit of IL-23 and thus it is possible that the C11.5 antibody will crossreact with IL-23.

Host Species: Mouse
Clone: C11.5
Isotype: IgG1
Species Reactivity: Human
Immunogen: CHO-expressed recombinant human IL-12 p70 heterodimer

Blocking Assay, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 (p40/p70)
Applications Blocking Assay, Flow Cytometry
Classification Monoclonal
Clone C11.5
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen CHO-expressed recombinant human IL-12 p70 heterodimer
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.