Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 (p40/p70) Rat anti-Mouse, NA/LE, Unlabeled, Clone: C17.8, BD

Catalog No. BDB554475 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554475 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554475 Supplier BD Biosciences Supplier No. 554475
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The C17.8 antibody reacts with the free p40 monomer of IL-12 as well as the p40 subunit complexed as homodimer or complexed with p35 as a p70 heterodimer. Crossreactivity to IL-23, which also contains the p40 subunit, has not been determined. The immunogen used to generate C17.8 hybridoma was recombinant mouse IL-12 p70.

Host Species: Rat
Clone: C17.8
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: Recombinant Mouse IL-12 p70

Functional Assay, Neutralization

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 (p40/p70)
Applications ELISA
Classification Monoclonal
Clone C17.8
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Immunogen Recombinant Mouse IL-12 p70
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.