Learn More
Description
The C11.5 antibody reacts with human IL-12 p40 monomer and p70 heterodimer, but not p35 monomer. The immunogen used to generate the C11.5 hybridoma was the CHO-expressed recombinant human IL-12 p70 heterodimer. p40 has also been described as a subunit of IL-23 and thus it is possible that the C11.5 antibody will crossreact with IL-23.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Intracellular Staining
Specifications
Specifications
| Antigen | IL-12 (p40/p70) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | C11.5 |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | CHO-expressed recombinant human IL-12 p70 heterodimer |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.