Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 (p40/p70) Mouse anti-Human, V450, Clone: C11.5, BD

Catalog No. BDB561380 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561380 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561380 Supplier BD Biosciences Supplier No. 561380
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C11.5 monoclonal antibody specifically binds to the human IL-12 p40 monomer and p70 heterodimer, but does not bind to the IL-12 p35 monomer. The immunogen used to generate the C11.5 hybridoma was the CHO-expressed recombinant human IL-12 p70 heterodimer. p40 has also been described as a subunit of IL-23 and thus it is possible that the C11.5 antibody crossreacts with IL-23.

The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 (p40/p70)
Applications Flow Cytometry
Classification Monoclonal
Clone C11.5
Conjugate Violet Fluor 450 (V450)
Description IL12B; IL-12B; CLMF; CLMF2; NKSF; NKSF2
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen CHO-expressed recombinant human IL-12 p70 heterodimer
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.