Learn More
IL-12 p70 Rat anti-Mouse, Unlabeled, Clone: 9A5, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 554658
Description
The 9A5 antibody reacts with the p70 subunit of mouse IL-12. The immunogen used to generate the 9A5 hybrodoma was CHO-expressed recombinant mouse IL-12 p70. This antibody is routinely tested by ELISA Capture. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 9A5
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: CHO-Expressed Recombinant Mouse IL-12 p70
ELISA

Specifications
IL-12 p70 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
CHO-Expressed Recombinant Mouse IL-12 p70 | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2b |
ELISA | |
9A5 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.