Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Rat anti-Human, Brilliant Violet 421, Clone: JES10-5A2, BD

Catalog No. BDB563580 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563580 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563580 Supplier BD Biosciences Supplier No. 563580
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The JES10-5A2 monoclonal antibody specifically binds to human interleukin-13, IL-13. IL-13 is produced by activated T cells, mast cells and NK cells. IL-13 regulates IgE production by B cells and can suppress the cytotoxic activity of macrophages and their production of inflammatory mediators. The immunogen used to produce the JES10-5A2 hybridoma was COS-expressed recombinant human IL-13. This is a neutralizing antibody.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Applications Flow Cytometry
Classification Monoclonal
Clone JES10-5A2
Conjugate Brilliant Violet 421
Description Interleukin-13; NC30; ALRH; BHR1; P600
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Human Recombinant IL-13
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.