Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13 Rat anti-Human, PE, Clone: JES10-5A2, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 340508
Description
Anti-Hu–IL-13, clone JES10-5A2, is derived from the mouse myeloma P3X63Ag8 fused with splenocytes from Lewis rats immunized with recombinant human IL-13 expressed in mammalian cells. Anti-Human Interleukin 13 (Anti-Hu–IL-13) recognizes a nonglycosylated protein of 132 amino acids with a molecular mass of 10kD.
Specifications
IL-13 | |
Monoclonal | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Human | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
JES10-5A2 | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human Recombinant IL-13 | |
50 Tests | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction