Learn More
IL-13 Rat anti-Human, PE, Clone: JES10-5A2, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 554571
Description
This antibody reacts with human IL-13. The immunogen used to produce the JES10-5A2 hybridoma was COS-expressed recombinant human IL-13. This is a neutralizing antibody.
Host Species: Rat
Clone: JES10-5A2
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human recombinant IL-13
Intracellular Staining

Specifications
IL-13 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human recombinant IL-13 | |
0.1 mg | |
Immunology | |
Human | |
IgG1 |
Flow Cytometry | |
JES10-5A2 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.