Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IL-17 RA/IL-17 R, Fluorescein, Clone: 133617, R&D Systems™
Specifications
Antigen | IL-17 RA/IL-17 R |
---|---|
Clone | 133617 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB177F025
|
R&D Systems
FAB177F025 |
25 Tests | N/A | N/A | N/A | ||||
FAB177F100
|
R&D Systems
FAB177F100 |
100 Tests | N/A | N/A | N/A | ||||
Specifications
IL-17 RA/IL-17 R | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 receptor A, IL17RA, IL-17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
IL17RA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human IL-17 R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-17 R, rmIL-17B R, or recombinant human IL-17B R is observed. In Western blots, no cross-reactivity with rmIL-17 R is observed |
133617 | |
Flow Cytometry | |
Fluorescein | |
Mouse | |
Human | |
Q96F46 | |
23765 | |
Mouse myeloma cell line NS0-derived recombinant human IL-17 R, Leu33-Trp320, Accession # Q96F46 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title