Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17A Rat anti-Mouse, Biotin, Clone: TC11-8H4.1, BD

Catalog No. BDB555067 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555067 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555067 Supplier BD Biosciences Supplier No. 555067
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TC11-8H4 antibody reacts with mouse IL-17A protein. The immunogen used to generate this hybrodoma was recombinant mouse IL-17 protein.

ELISA, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17A
Applications ELISA
Classification Monoclonal
Clone TC11-8H4.1
Concentration 0.5mg/mL
Conjugate Biotin
Description IL-17; Il17; IL-17A; Il17a; CTLA-8; Ctla8; Interleukin-17A; interleukin 17A
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant mouse IL-17
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.