Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-17A, BV786 ; Clone: TC11-18H10; BD

Catalog No. BDB564171 Shop All BD Biosciences Products
Change view
Click to view available options
Conjugate:
Brilliant Violet 786
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
BDB564171 Brilliant Violet 786
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564171 Supplier BD Biosciences Supplier No. 564171
Add to Cart
Add to Cart

Applications:

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17A
Applications Flow Cytometry
Classification Monoclonal
Clone TC11-18H10
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Host Species Rat
Immunogen Recombinant Mouse IL-17A Protein
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.