Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17A Mouse anti-Human, V450, Clone: N49-653, BD

Catalog No. BDB560610 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560610 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560610 Supplier BD Biosciences Supplier No. 560610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human IL-17A, also known as IL-17, is a proinflammatory cytokine that is encoded by the IL17A gene in chromosome 6. IL-17A is produced as a disulfide-linked homodimer comprised of two mature 136-amino acid polypeptides. It is a member of the IL-17 family

Host Species: Mouse
Clone: N49-653
Isotype: IgG1 κ
Species Reactivity: Human

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17A
Applications Flow Cytometry
Classification Monoclonal
Clone N49-653
Conjugate Violet Fluor 450 (V450)
Description IL-17; CTLA8; Cytotoxic T-lymphocyte-associated serine esterase 8
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IL-17A Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.