Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IL-17F, APC, Clone: 197301, R&D Systems™
Specifications
Antigen | IL-17F |
---|---|
Clone | 197301 |
Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
IC13351A025
|
R&D Systems
IC13351A025 |
25 Tests | N/A | N/A | N/A | ||||
IC13351A100
|
R&D Systems
IC13351A100 |
100 Tests | N/A | N/A | N/A | ||||
Specifications
IL-17F | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Cytokine ML-1, IL17F, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24 | |
IL17F | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human IL-17F in direct ELISAs. Does not cross-react with recombinant human IL-17A, B, C, D, or E. |
197301 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
Q96PD4 | |
112744 | |
E. coli-derived recombinant human IL-17F, Arg31-Gln163, Accession # Q96PD4 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title