Learn More
IL-1b Hamster anti-Mouse, NA/LE, Unlabeled, Clone: B122, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 557539
Description
The B122 antibody recognizes the precursor and mature secreted forms of the mouse IL-1β protein. B122 neutralizes the biological activity of mouse IL-1β, but not the biological activities of mouse IL-1α nor human IL-1β. B122 does not recognize mouse IL-2, IL-4, IL-6, IL-10, TNF or IFN-γ. This antibody also recognizes rat IL-1β. The immunogen used to generate the B122 hybridoma was a purified, mature form of the mouse IL-1β protein.
Host Species: Hamster
Clone: B122
Isotype: IgG1, λ
Cross Reactivity: Mouse
Functional Assay, Neutralization

Specifications
IL-1b | |
Monoclonal | |
1.0mg/mL | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Mouse IL-1β Protein | |
0.25 mg | |
Immunology | |
Mouse | |
IgG λ1 |
ELISA | |
B122 | |
Unconjugated | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.