Learn More
Description
Specifically detects IL-2 Clone: JES6-5H4 in Mouse samples, and it is validated for Flow Cytometry
Specifications
Specifications
| Antigen | IL-2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | JES6-5H4 |
| Conjugate | Biotin |
| Formulation | Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. |
| Gene Accession No. | P04351 |
| Gene Symbols | Il2, Il-2 |
| Host Species | Rat |
| Immunogen | Proprietary immunogen |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.