Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 Mouse anti-Human, Unlabeled, Clone: 5344.111, BD

Catalog No. BDB555051 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555051 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555051 Supplier BD Biosciences Supplier No. 555051
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 5344.111 antibody reacts with human interleukin-2 (IL-2). The immunogen used to generate the 5344.111 hybridoma was affinity-purified, recombinant human IL-2 protein.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications ELISA
Classification Monoclonal
Clone 5344.111
Concentration 0.5mg/mL
Conjugate Unconjugated
Description IL2; Interleukin-2; T-cell growth factor; TCGF
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IL-2 protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.