Learn More
IL-2 Rat anti-Mouse, Unlabeled, Clone: JES6-5H4, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 554425
Description
The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2) and has been reported to be neutralizing.
Host Species: Rat
Clone: JES6-5H4
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Mouse IL-2 Recombinant Protein
Blocking Assay, Immunoprecipitation, Intracellular Staining

Specifications
IL-2 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse IL-2 Recombinant Protein | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b |
Blocking Assay, Flow Cytometry | |
JES6-5H4 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.