Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 Rat anti-Mouse, NA/LE, Unlabeled, Clone: S4B6, BD

Catalog No. BDB554375 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554375 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554375 Supplier BD Biosciences Supplier No. 554375
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The S4B6 antibody reacts with mouse interleukin-2 (IL-2). The immunogen used to generate the S4B6 hyridoma was a T cell clone supernatant. This is a neutralizing antibody.

Host Species: Rat
Clone: S4B6
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: T cell clone supernatant

Functional Assay, Neutralization

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications Neutralization
Classification Monoclonal
Clone S4B6
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing protein stabilizer, no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Immunogen T cell clone supernatant
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.