Learn More
Description
Anti-Human Interleukin-2 (Anti-Hu++IL-2) recognizes a 15- to 18-kilodalton (kd) glycoprotein. IL-2 is primarily synthesized by activated CD4+ T-helper lymphocytes (TH0 and TH1) and also by some CD8+ T lymphocytes. Upon polyclonal or antigenic stimulation, naive T lymphocytes express mainly IL-2. Memory TH1 lymphocytes synthesize IL-2 which mediates (along with other TH1 cytokines) delayed-type hypersensitivity responses. Upon activation, IL-2 is produced and binds to its receptor, resulting in proliferation and differentiation of T, B, and natural killer (NK) lymphocytes. The presence of IL-2 enhances NK-lymphocyte cytotoxicity. IL-2 is also a potent activator of monocytes. Anti-Hu++IL-2, clone 5344.111, is derived from a fusion of Sp2/0 myeloma cells with splenocytes from BALB/c mice immunized with recombinant human IL-2. Anti-Hu++IL-2 is composed of mouse IgG1 heavy chains and kappa light chains.
Host Species: Mouse
Clone: 5344.111
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human IL-2 protein
Flow Cytometry, Intracellular Staining
Specifications
Specifications
| Antigen | IL-2 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5344.111 |
| Concentration | 1.6μg/mL |
| Conjugate | PE |
| Formulation | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human IL-2 protein |
| Quantity | 50 Tests |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.