Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 Rα Mouse anti-Human, Clone: 7G7/B6, MilliporeSigma™

Catalog No. 05170MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-170-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-170-MI Supplier MilliporeSigma Supplier No. 05170
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-IL-2 Rα Antibody, clone 7G7/B6 is validated for use in DB, IC for the detection of IL-2 Rα.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2 Rα
Applications Dot Blot, Immunocytochemistry
Classification Monoclonal
Clone 7G7/B6
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative.
Gene Accession No. NM_000417.1
Gene Symbols IL2RA, CD25, IL2-RA, IL-2-RA, OTTHUMP00000044870, TCGFR, IL2R, IDDM10, p55
Host Species Mouse
Immunogen Whole cells expressing human IL-2 Receptor α
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Lyophilized: 2 years at −20°C; Rehydrated: 6 months at −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.