Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21 Mouse anti-Human, Alexa Fluor 647, Clone: 3A3-N2.1, BD

Catalog No. BDB560493 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560493 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560493 Supplier BD Biosciences Supplier No. 560493
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human Interleukin-21 (IL-21) is a member of the type I cytokine family that is encoded by a gene resident on chromosome 4. The mature form of human IL-21 is a 131 amino acid protein. IL-21 is produced by activated NKT and multiple CD4+ T cell subsets including effector memory and central memory CD4+ T cells and differentiated T helper cell subsets polarized towards Th17 cell and T follicular helper (Tfh) phenotypes. IL-21 plays important protective roles in the regulation of hematopoiesis and both innate and adaptive immune responses and adverse roles in promoting autoimmunity. IL-21 costimulates the proliferation and differentiation of CD4+ T cells. It enhances the proliferation of and cytotoxicity mediated by natural killer (NK) cells and CD8+ T cells. IL-21 costimulates B cell proliferation and differentiation into plasma cells producing immunoglobulins with IgG isotypes. IL-21 can also regulate the functions of dendritic cells and other myeloid cells. IL-21 exerts its biological activities by binding to and activating the Janus activating kinases (JAK1 and JAK3) and signal transducers and activators of transcription (STAT1, STAT3, STA5a and STAT5b) signaling pathways through the IL-21 receptor (IL-21R)
complex. The IL-21R complex is comprised of the IL-21R α subunit and the common cytokine receptor gamma subunit (γ c; CD132). The monoclonal 3A3-N2 antibody specifically binds to human IL-21.

Host Species: Mouse
Clone: 3A3-N2.1
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-21
Applications Flow Cytometry
Classification Monoclonal
Clone 3A3-N2.1
Conjugate Alexa Fluor 647
Description Interleukin-21, IL21, Za11
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.