Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21 Mouse anti-Human, Biotin, Clone: I76-539, BD

Catalog No. BDB558502 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558502 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558502 Supplier BD Biosciences Supplier No. 558502
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The I76-539 antibody reacts with interleukin-21 (IL-21), an ∽15kDa member of the type-I-cytokine family of proteins. IL-21 is expressed primarily by CD4+ T cells and has a broad range of immunoregulatory functions on B cells, CD8+ T cells, NK cells and dendritic cells. The effects of IL-21 include promotion of antibody production, NK cell differentiation, proliferation and differentiation of cells of monocyte and granulocyte lineages, costimulation of T cells and augmentation of antitumor activity of CD8+ T cells. It has been speculated that IL-21, through its varies positiveand negative regulatory effects on cell-mediated and humoral immunity, may play a role in autoimmuninty, allergy, asthma and tumor regression.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-21
Applications ELISA
Classification Monoclonal
Clone I76-539
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IL-21 Recombinant Protein
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.